EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Removal of parabens and their chlorinated by-products by periphyton: influence of light and temperature.
PMID 28032288
Abstract
The extensive use of parabens as preservatives in food and pharmaceuticals and personal care products results in frequent detection of their residuals in aquatic environment. In this work, the adsorption and removal of four parabens (methyl-, ethyl-, propyl-, and butyl-paraben) and two chlorinated methyl-parabens (CMPs) by periphyton were studied. Characteristics of the periphyton were identified to explore the possible relationship between paraben removal and periphyton properties. Results showed that linear adsorption coefficients (K