The Journal of surgical research
Bradykinin stimulates protein kinase D-mediated colonic myofibroblast migration via cyclooxygenase-2 and heat shock protein 27.
PMID 28032559
Inflammatory bowel disease is characterized by episodic intestinal injury and repair. Myofibroblasts are gastrointestinal tract stromal cells that regulate the reparative process and are known targets of inflammatory mediators including bradykinin (BK). However, the mechanisms through which inflammation regulates myofibroblast-induced wound healing remain incompletely understood. Here, we demonstrate, for the first time, that BK stimulates myofibroblast migration through protein kinase D (PKD)-mediated activation of the cyclooxygenase-2 (COX-2) and heat shock protein 27 (Hsp27) pathways. CCD-18Co is a human colonic myofibroblast cell line used from passages 8 to 14. An inxa0vitro scratch assay assessed the effect of BK (100xa0nM) on myofibroblast migration over 24xa0h in the presence or absence of several inhibitors (CID755673 [10xa0μM] and NS398 [10xa0μM]). Hsp27 small interfering RNA evaluated the effect of Hsp27 on colonic myofibroblast migration. Antibodies to pPKD, pHsp27, and COX-2 evaluated expression levels by Western blot. BK stimulated myofibroblast migration over 24xa0h. BK also led to rapid and sustained phosphorylation of PKD at Ser-916, rapid phosphorylation of Hsp27 at Ser-82, and increased COX-2 expression over 4xa0h. BK-mediated COX-2 expression and Hsp27 phosphorylation were both inhibited by the PKD inhibitor CID755673. Similarly, BK-induced myofibroblast migration was significantly inhibited by CID755673 (Pxa0
Related Materials
Product #
Molecular Formula
Add to Cart
EHU112501 MISSION® esiRNA, esiRNA human HSPB1 (esiRNA1)