Evaluation of a hollow fiber supported liquid membrane device as a chemical surrogate for the measurements of zinc (II) bioavailability using two microalgae strains as biological references.
PMID 28033574
The environmental bioavailability of zinc (II), i.e., the uptake of the element by an organism, was determined using two microalgae species, Scenedesmus acutus and Pseudokirchneriella subcapitata, and estimated using hollow fiber supported liquid membrane (HF-SLM) device as the chemical surrogate. Several experimental conditions were studied including the presence of organic matter, inorganic anions and concomitant cations and pH. The results show strong positive correlation coefficients between the responses given by the HF-SLM and the microalgae species (rxa0=xa00.900 for S. acutus and rxa0=xa00.876 for P. subcapitata) in multivariate environments (changes in pH, calcium, humic and citrate concentrations). The maximum amount of zinc (II) retained by the HF-SLM (4.7xa0×xa010(-8)xa0mol/cm(2)) was higher than those for P. subcapitata and S. acutus (9.4xa0×xa010(-11)xa0mol/cm(2) and 6.2xa0×xa010(-11)xa0mol/cm(2), respectively). The variation in pH (pH 5.5-9) was the variable with the greatest effect on zinc internalization in all systems, increasing approximately 2.5 times for P. subcapitata and 5.5 times for S. acutus respect to pHxa0=xa05.5, while the presence of humic acids did not affect the response. The species' concentration analysis of the experimental design at pHxa0=xa05.5 indicated that the amount of internalized zinc (II) by the HF-SLM and both microalgae species is strongly dependent on the free zinc concentration (rxa0=xa00.910 for the HF-SLM, rxa0=xa00.922 for S. acutus and rxa0=xa00.954 for P. subcapitata); however, at pHxa0=xa09.0, the amount of internalized zinc (II) is strongly dependent on the sum of free zinc and labile species (rxa0=xa00.912 for the HF-SLM, rxa0=xa00.947 for S. acutus and rxa0=xa00.900 for P. subcapitata). The presence of inorganic ligands (chloride, sulfate, phosphate, carbonate, and nitrate) and metal ions (cobalt (II), copper (II), nickel (II), chromium (VI), lead (II) and cadmium (II)) produced different behaviors both in the chemical surrogate and the biological references. The results showed that the synthetic device can mimic biological uptake in the presence of humic acids, nitrate, sulfate, and phosphate, and pH within the range 5.5-9 when S. acutus was used as the biological reference, considering the simultaneous contribution of the Zn(2+) and ZnOH(+) labile species depending on the chemical composition of the medium.
Related Materials
Product #
Molecular Formula
Add to Cart
Hexylbenzene, 97%