EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
A novel phloroglucinol and two new phenolic glycosides from Psidium littorale.
PMID 28033721
Abstract
One novel phloroglucinol, psidosone A (1), and two new phenolic glycosides, psidoside A (2), and psidoside B (3), together with nine known phenol compounds (4-12), were isolated from the fruits of Psidium littorale Raddi. Their structures were elucidated using data obtained from MS,