EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
High expression of organic cation transporter 3 in human BAT-like adipocytes. Implications for extraneuronal norepinephrine uptake.
PMID 28034777
Abstract
Brown adipose tissue (BAT) is activated by extracellular norepinephrine (NE) released by the sympathetic nervous system. The extracellular concentration of NE is additionally regulated by the disappearance/degradation of NE. Recent studies have introduced the organic cation transporter 3 (OCT3) as a possible contributor in the regulation of NE in adipose tissue. In the present study we set out to investigate the presence of OCT3 in human neck adipose tissue (AT), which is the primary localization of BAT in humans. Moreover, we wanted to assess the possible function and correlation of the transporter with known markers of thermogenic function, e.g. UCP1. When examining neck AT biopsies from 57 individuals we found that OCT3 was expressed at 2.5 ± 0.16 fold higher level in the deep-neck AT compared with subcutaneous AT. UCP1 was found extensively expressed in the deep-neck AT depot and the correlation between UCP1 and OCT3 within the deep-neck AT was found highly significant (r