EMAIL THIS PAGE TO A FRIEND
Biomaterials
Decellularized adipose tissue microcarriers as a dynamic culture platform for human adipose-derived stem/stromal cell expansion.
PMID 28038353
Abstract
With the goal of designing a clinically-relevant expansion strategy for human adipose-derived stem/stromal cells (ASCs), methods were developed to synthesize porous microcarriers derived purely from human decellularized adipose tissue (DAT). An electrospraying approach was applied to generate spherical DAT microcarriers with an average diameter of 428 ± 41 μm, which were soft, compliant, and stable in long-term culture without chemical crosslinking. Human ASCs demonstrated enhanced proliferation on the DAT microcarriers relative to commercially-sourced Cultispher-S microcarriers within a spinner culture system over 1 month. ASC immunophenotype was maintained post expansion, with a trend for reduced expression of the cell adhesion receptors CD73, CD105, and CD29 under dynamic conditions. Upregulation of the early lineage-specific genes PPARγ, LPL, and COMP was observed in the ASCs expanded on the DAT microcarriers, but the cells retained their multilineage differentiation capacity. Comparison of adipogenic and osteogenic differentiation in 2-D cultures prepared with ASCs pre-expanded on the DAT microcarriers or Cultispher-S microcarriers revealed similar adipogenic and enhanced osteogenic marker expression in the DAT microcarrier group, which had undergone a higher population fold change. Further, histological staining results suggested a more homogeneous differentiation response in the ASCs expanded on the DAT microcarriers as compared to either Cultispher-S microcarriers or tissue culture polystyrene. A pilot chondrogenesis study revealed higher levels of chondrogenic gene and protein expression in the ASCs expanded on the DAT microcarriers relative to all other groups, including the baseline controls. Overall, this study demonstrates the promise of applying dynamic culture with tissue-specific DAT microcarriers as a means of deriving regenerative cell populations.