EMAIL THIS PAGE TO A FRIEND
Oncotarget
Nuclear overexpression of the overexpressed in lung cancer 1 predicts worse prognosis in gastric adenocarcinoma.
PMID 28038462
Abstract
We have performed this retrospective study to elucidate whether elevated expression of the overexpressed in lung cancer 1 (OLC1) was related to the clinicopathological parameters and prognosis of patients with gastric adenocarcinoma. Additionally, different effects of various subcellular OLC1 expression on gastric adeno-carcinogenesis were focused on in our study. Both overall and subcellular expression of OLC1 was evaluated by immunohistochemistry(IHC) via tissue microarrays from total 393 samples. The Kaplan-Meier method and Cox's proportional hazard model were exerted to further explore the correlation between OLC1 and prognosis. Total overexpression of OLC1 was significantly associated with stage (P = 0.004) and differentiation (P = 0.009), and only the strong total expression could predict a poor prognosis (HR = 1.31, P = 0.04). There were significant associations found between nuclear overexpression and tumor invasion depth(P = 0.002), lymph node (P < 0.001), stage (P = 0.004), differentiation (P < 0.001) and smoking history (P = 0.045). Furthermore, over-expressed nuclear OLC1 protein could be an independent risk factor for gastric adenocarcinoma (univariate: HR = 1.43, P = 0.003; multivariate: HR = 1.39, P = 0.011). In general, both total and nuclear overexpression of OLC1 could be the signs of gastric adeno-carcinogenesis, which might be served as the biomarkers for diagnosis at an early stage, even at the onset of tumorigenesis. Rather than the total expression, nuclear overexpression of OLC1 was correlated with most clinicopathological parameters and could predict a poor overall survival as an independent factor for prognosis, which made it a more effective and sensitive biomarker for gastric adenocarcinoma.