Drug-dependent functionalization of wild-type and mutant p53 in cisplatin-resistant human ovarian tumor cells.
PMID 28038466
Cisplatin (cis-Pt) resistance in tumor cells from p53 dysfunction is a significant clinical problem. Although mutation can inhibit p53 function, >60% of p53 mutants retain normal function according to literature reports. Therefore, we examined the status of p53 in cisplatin-resistant ovarian tumor models and its functional response to cis-Pt and the mechanistically-distinct non-cross-resistant oxaliplatin (oxali-Pt). Relative to sensitive A2780 cells harboring wild-type p53, the 2780CP/Cl-16, OVCAR-10, Hey and OVCA-433 cell lines were 10- to 30-fold resistant to cis-Pt, but was substantially circumvented by oxali-Pt. Mutant p53 in 2780CP/Cl-16 (p53V172F) and OVCAR-10 (p53V172F and p53G266R) cells, predicted as non-functional in p53 database, displayed attenuated response to cis-Pt, as did the polymorphic p53P72R (functionally equivalent to wild-type p53) in HEY and OVCA-433 cell lines. However, p53 was robustly activated by oxali-Pt in all cell lines, with resultant drug potency confirmed as p53-dependent by p53 knockout using CRISPR/Cas9 system. This p53 activation by oxali-Pt was associated with phosphorylation at Ser20 by MEK1/2 based on inhibitor and kinase studies. Cis-Pt, however, failed to phosphorylate Ser20 due to downregulated Chk2, and its clinical impact validated by reduced overall survival of ovarian cancer patients according to TCGA database. In conclusion, cis-Pt resistance occurs in both wild-type and mutant p53 ovarian cancer cells, but is associated with loss of Ser20 phosphorylation. However, these mutant p53, like polymorphic p53, are functional and activated by oxali-Pt-induced Ser20 phosphorylation. Thus, the potential exists for repurposing oxali-Pt or similar drugs against refractory cancers harboring wild-type or specific mutant p53.