EMAIL THIS PAGE TO A FRIEND
Oncotarget
Inhibition of ALDH2 by O-GlcNAcylation contributes to the hyperglycemic exacerbation of myocardial ischemia/reperfusion injury.
PMID 28038474
Abstract
Although hyperglycemia is causally related to adverse outcomes after myocardial ischemia/reperfusion (I/R), the underlying mechanisms are largely unknown. Here, we investigated whether excessive O-linked-N-acetylglucosamine (O-GlcNAc) modification of acetaldehyde dehydrogenase 2 (ALDH2), an important cardioprotective enzyme, was a mechanism for the hyperglycemic exacerbation of myocardial I/R injury. Both acute hyperglycemia (AHG) and diabetes (DM)-induced chronic hyperglycemia increased cardiac dysfunction, infarct size and apoptosis index compared with normal saline (NS)+I/R rats (P<0.05). ALDH2 O-GlcNAc modification was increased whereas its activity was decreased in AHG+I/R and DM+I/R rats. High glucose (HG, 30mmol/L) markedly increased ALDH2 O-GlcNAc modification compared with Con group (5mmol/L) (P<0.05). ALDH2 O-GlcNAc modification was increased by 62.9% in Con+PUGNAc group whereas it was decreased by 44.1% in Con+DON group compared with Con group (P<0.05). Accordingly, ALDH2 activity was decreased by 18.1% in Con+PUGNAc group whereas it was increased by 17.9% in Con+DON group. Moreover, DON decreased levels of 4-hydroxy-2-nonenal (4-HNE), aldehydes, protein carbonyl accumulation and apoptosis index compared with HG+H/R group (P<0.05). Alda-1, a specific activator of ALDH2, significantly decreased ALDH2 O-GlcNAc modification and improved infarct size, apoptosis index and cardiac dysfunction induced by I/R combined with hyperglycemia. These findings demonstrate that ALDH2 O-GlcNAc modification is a key mechanism for the hyperglycemic exacerbation of myocardial I/R injury and Alda-1 has therapeutic potential for inducing cardioprotection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0462
Alda-1, ≥98% (HPLC)
C15H11Cl2NO3
A7229
PUGNAc, ≥95% (HPLC)
C15H19N3O7