EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Inhibition of Ubc13-mediated Ubiquitination by GPS2 Regulates Multiple Stages of B Cell Development.
PMID 28039360
Abstract
Non-proteolytic ubiquitin signaling mediated by Lys