EMAIL THIS PAGE TO A FRIEND
Aging
Sodium fluoride (NaF) induces the splenic apoptosis via endoplasmic reticulum (ER) stress pathway
PMID 28039491
Abstract
At present, there are no reports on the relationship between fluoride-induced apoptosis and endoplasmic reticulum (ER) stress (ER stress) in the spleen of human and animals