EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Hydroxyl radical generation by dissociation of water molecules during 1.65 MHz frequency ultrasound irradiation under aerobic conditions.
PMID 28040432
Abstract
The dissociation of water molecules by ultrasound irradiation under aerobic conditions was demonstrated experimentally. To be able to detect the dissociation of water molecules, we performed the ultrasound irradiation of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56207
1-Hydroxypiperidine, ≥96.0% (GC)
C5H11NO