EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
Androgen receptor regulation by histone methyltransferase Suppressor of variegation 3-9 homolog 2 and Melanoma antigen-A11.
PMID 28042025
Abstract
Androgen receptor (AR) transcriptional activity depends on interactions between the AR NH