EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Adaptive responses of heart and skeletal muscle to spermine oxidase overexpression: Evaluation of a new transgenic mouse model.
PMID 28043891
Abstract
Spermine oxidase oxidizes spermine to produce H