EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Calibration of the freshwater mussel, Elliptio complanata, for quantitative biomonitoring of hexachlorobenzene and octachlorostyrene in aquatic systems.
PMID 2804398
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCO-001
Octachlorostyrene, vial of 1 g, analytical standard
C8Cl8