EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Priming of endothelial colony-forming cells in a mesenchymal niche improves engraftment and vasculogenic potential by initiating mesenchymal transition orchestrated by NOTCH signaling.
PMID 28045376
Abstract
The prospect of using endothelial progenitors is currently hampered by their low engraftment upon transplantation. We report that mesenchymal stem/stromal cells (MSCs), independent of source and age, improve the engraftment of endothelial colony forming cells (ECFCs). MSC coculture altered ECFC appearance to an elongated mesenchymal morphology with reduced proliferation. ECFC primed via MSC contact had reduced self-renewal potential, but improved capacity to form tube structures in vitro and engraftment in vivo Primed ECFCs displayed major differences in transcriptome compared to ECFCs never exposed to MSCs, affecting genes involved in the cell cycle, up-regulating of genes influencing mesenchymal transition, adhesion, extracellular matrix. Inhibition of NOTCH signaling, a potential upstream regulator of mesenchymal transition, in large part modulated this gene expression pattern and functionally reversed the mesenchymal morphology of ECFCs. The collective results showed that primed ECFCs survive better and undergo a mesenchymal transition that is dependent on NOTCH signaling, resulting in significantly increased vasculogenic potential.-Shafiee, A., Patel, J., Wong, H. Y., Donovan, P., Hutmacher, D. W., Fisk, N. M., Khosrotehrani, K. Priming of endothelial colony-forming cells in a mesenchymal niche improves engraftment and vasculogenic potential by initiating mesenchymal transition orchestrated by NOTCH signaling.