EMAIL THIS PAGE TO A FRIEND
AMB Express
Microbial biomass in compost during colonization of Agaricus bisporus.
PMID 28050852
Abstract
Agaricus bisporus mushrooms are commercially produced on a microbe rich compost. Here, fungal and bacterial biomass was quantified in compost with and without colonization by A. bisporus. Chitin content, indicative of total fungal biomass, increased during a 26-day period from 576 to 779 nmol N-acetylglucosamine g
Related Materials