EMAIL THIS PAGE TO A FRIEND
Scientific reports
The hypoparathyroidism-associated mutation in Drosophila Gcm compromises protein stability and glial cell formation.
PMID 28051179
Abstract
Differentiated neurons and glia are acquired from immature precursors via transcriptional controls exerted by factors such as proteins in the family of Glial Cells Missing (Gcm). Mammalian Gcm proteins mediate neural stem cell induction, placenta and parathyroid development, whereas Drosophila Gcm proteins act as a key switch to determine neuronal and glial cell fates and regulate hemocyte development. The present study reports a hypoparathyroidism-associated mutation R59L that alters Drosophila Gcm (Gcm) protein stability, rendering it unstable, and hyperubiquitinated via the ubiquitin-proteasome system (UPS). Gcm