EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Glutamine attenuates obstructive cholestasis in rats via farnesoid X receptor-mediated regulation of Bsep and Mrp2.
PMID 28051334
Abstract
To investigate the protective effect of glutamine (Gln) against obstructive cholestasis in association with farnesoid X receptor (FXR) activation, an obstructive cholestasis model was established in male Sprague-Dawley rats by bile duct ligation (BDL). Serum biomarkers and hematoxylin plus eosin staining were used to identify the degree of hepatic injury in the rats with obstructive cholestasis after Gln treatment. Immunohistochemistry, real-time PCR, Western blot, cultured primary rat hepatocytes with FXR knockdown, and dual-luciferase reporter assay were performed to elucidate the mechanisms underlying Gln hepatoprotection. We found that Gln treatment protected against obstructive cholestasis induced by BDL through reducing hepatocyte injury. Upregulation of the hepatic efflux transporters small heterodimer partner (Shp), bile salt export pump (Bsep), and multidrug resistance-associated protein 2 (Mrp2), and inhibition of the hepatic uptake transporter Na