EMAIL THIS PAGE TO A FRIEND
Cell reports
EBI2 Expression and Function: Robust in Memory Lymphocytes and Increased by Natalizumab in Multiple Sclerosis.
PMID 28052250
Abstract
The interaction between oxysterols and the G protein-coupled receptor Epstein-Barr virus-induced gene 2 (EBI2) fine-tunes immune cell migration, a mechanism efficiently targeted by several disease-modifying treatments developed to treat multiple sclerosis (MS), such as natalizumab. We previously showed that memory CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1981 NIBR189, ≥98% (HPLC)
C21H21BrN2O3