EMAIL THIS PAGE TO A FRIEND
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
An HDAC Inhibitor, Entinostat/MS-275, Partially Prevents Delayed Cranial Suture Closure in Heterozygous Runx2 Null Mice.
PMID 28052439
Abstract
Cleidocranial dysplasia (CCD) is an autosomal dominant skeletal disorder caused by mutations in RUNX2, coding a key transcription factor of early osteogenesis. CCD patients suffer from developmental defects in cranial bones. Despite numerous investigations and clinical approaches, no therapeutic strategy has been suggested to prevent CCD. Here, we show that fetal administration of Entinostat/MS-275, a class I histone deacetylase (HDAC)-specific inhibitor, partially prevents delayed closure of cranial sutures in Runx2
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3882
Alizarin-3-methyliminodiacetic acid
C19H15NO8
EPS002
MS-275, A HDAC1 and HDAC3 inhibitor
C21H20N4O3