EMAIL THIS PAGE TO A FRIEND
Saudi journal of biological sciences
Comparison of phenotypic and PCR methods for detection of carbapenemases production by Enterobacteriaceae.
PMID 28053586
Abstract
Dissemination of carbapenem resistance via Enterobacteriaceae, particularly among