EMAIL THIS PAGE TO A FRIEND
Basic & clinical pharmacology & toxicology
Opposite Modulatory Effects of Selective and Non-Selective Cyclooxygenase Inhibition on Cardiovascular and Autonomic Consequences of Cyclosporine in Female Rats.
PMID 28054752
Abstract
Non-steroidal anti-inflammatory drugs (NSAIDs) and the immunosuppressant drug cyclosporine (CSA) are concurrently administered in arthritis. Here, we investigated whether diclofenac (non-selective inhibitor of cyclooxygenase-1 and 2, COX-1/COX-2), celecoxib (selective COX-2 inhibitor) or SC560 (selective COX-1 inhibitor) interact variably with haemodynamic effects of CSA in conscious female rats. Changes caused by CSA (10 mg/kg i.v.) in blood pressure (BP), heart rate (HR), HR variability (HRV) and myocardial contractility were assessed in the absence and presence of individual NSAIDs (10 mg/kg each). Compared with vehicle values, CSA caused immediate and sustained (i) increases in BP and decreases in pulse pressure index (PPI) and HR, (ii) elevations in left ventricular (LV) contractility (dP/dt
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D35201
N,N′-Dibenzylethylenediamine, 97%
C16H20N2