EMAIL THIS PAGE TO A FRIEND
ACS synthetic biology
Orthogonal Genetic Regulation in Human Cells Using Chemically Induced CRISPR/Cas9 Activators.
PMID 28054767
Abstract
The concerted action of multiple genes in a time-dependent manner controls complex cellular phenotypes, yet the temporal regulation of gene expressions is restricted on a single-gene level, which limits our ability to control higher-order gene networks and understand the consequences of multiplex genetic perturbations. Here we developed a system for temporal regulation of multiple genes. This system combines the simplicity of CRISPR/Cas9 activators for orthogonal targeting of multiple genes and the orthogonality of chemically induced dimerizing (CID) proteins for temporal control of CRISPR/Cas9 activator function. In human cells, these transcription activators exerted simultaneous activation of multiple genes and orthogonal regulation of different genes in a ligand-dependent manner with minimal background. We envision that our system will enable the perturbation of higher-order gene networks with high temporal resolution and accelerate our understanding of gene-gene interactions in a complex biological setting.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1959 GA3-AM, ≥95% (HPLC)
C22H26O8