EMAIL THIS PAGE TO A FRIEND
eLife
APP modulates KCC2 expression and function in hippocampal GABAergic inhibition.
PMID 28054918
Abstract
Amyloid precursor protein (APP) is enriched at the synapse, but its synaptic function is still poorly understood. We previously showed that GABAergic short-term plasticity is impaired in
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2172
CLP290, ≥98% (HPLC)
C19H21FN4O3S