EMAIL THIS PAGE TO A FRIEND
Cell communication and signaling : CCS
Aldose reductase mediates endothelial cell dysfunction induced by high uric acid concentrations.
PMID 28057038
Abstract
Uric acid (UA) is an antioxidant found in human serum. However, high UA levels may also have pro-oxidant functions. According to previous research, aldose reductase (AR) plays a vital role in the oxidative stress-related complications of diabetes. We sought to determine the mechanism by which UA becomes deleterious at high concentrations as well as the effect of AR in this process. Endothelial cells were divided into three groups cultured without UA or with 300 μM or 600 μM UA. The levels of total reactive oxygen species (ROS), of four ROS components, and of NO and NOX4 expression were measured. Changes in the above molecules were detected upon inhibiting NOX4 or AR, and serum H Increased AR expression in high UA-treated endothelial cells enhanced ROS production by activating NADPH oxidase. These effects were blocked by the AR inhibitor epalrestat. 300 μM UA decreased the levels of the three major reactive oxygen species (ROS) components: O Our findings indicate that inhibiting AR or degrading H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0046
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)
C13H10N2O