EMAIL THIS PAGE TO A FRIEND
Scientific reports
CNTs-Modified Nb
PMID 28059123
Abstract
Novel semiconductor photocatalysts have been the research focus and received much attention in recent years. The key issues for novel semiconductor photocatalysts are to effectively harvest solar energy and enhance the separation efficiency of the electron-hole pairs. In this work, novel Nb