EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Cubane-like tetranuclear Cu(ii) complexes bearing a Cu
PMID 28059406
Abstract
In the present work, two new copper complexes 3a and 3b with a Cu
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447706
5-Chlorosalicylaldehyde, 98%
C7H5ClO2