EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Aspirin and (or) omega-3 polyunsaturated fatty acids protect against corticohippocampal neurodegeneration and downregulate lipoxin A
PMID 28060522
Abstract
There is evidence for a relationship between inflammation and seizures because epilepsy can be caused by or result in inflammation. This study aimed to investigate the effect of aspirin and (or) omega-3 polyunsaturated fatty acids (PUFAs) on seizure activity and neurodegeneration in pentylenetetrazole (PTZ)-kindled rats focusing on their effect on corticohippocampal production of lipoxin A