EMAIL THIS PAGE TO A FRIEND
Oncotarget
B7-H4 promotes tumor growth and metastatic progression in lung cancer by impacting cell proliferation and survival.
PMID 28061481
Abstract
Aberrant expression of B7-H4 occurs across a broad spectrum of human cancers. The aim of this study was to investigate the key role of B7-H4 during tumorigenesis and metastasis of human lung cancer. Our data showed that the shRNA-mediated disruption of B7-H4 markedly inhibited tumor cell proliferation, invasion and migration, increased cell apoptosis and arrested cell cycle at G0/G1. These changes were accompanied by a marked increase in Bax and caspase-3/caspase-8, but a decrease in Bcl-2, cyclinD1 and activation of AKT. In addition, our shRNA-mediated disruption of B7-H4 led to a marked decrease in tumor growth in the immune-compromised mice. Importantly, B7-H4 was expressed in 53.33% of lung carcinomas from our patient cohort (n = 90), but not in any of adjacent non-cancerous tissues, according to our IHC analyses. In particular, B7-H4 expression appeared to be associated with lymph node metastasis (P = 0.008) and TNM stage (P = 0.012). Taken together, our study demonstrates a strong promoting role of B7-H4 in lung tumor growth, progression and metastasis, and supports its potential as a therapeutic target for the treatment of the disease.