EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Regulation of Chemokine Signal Integration by Activator of G-Protein Signaling 4 (AGS4).
PMID 28062526
Abstract
Activator of G-protein signaling 4 (AGS4)/G-protein signaling modulator 3 (Gpsm3) contains three G-protein regulatory (GPR) motifs, each of which can bind G