EMAIL THIS PAGE TO A FRIEND
Water research
Role of calcium ions on the removal of haloacetic acids from swimming pool water by nanofiltration: mechanisms and implications.
PMID 28063295
Abstract
We investigated the removal of haloacetic acids (HAAs) from swimming pool waters (SPWs) by two nanofiltration membranes NF270 and NF90. The strong matrix effect (particularly by Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330574
Indophenol Blue, Dye content 60 %
C18H16N2O