EMAIL THIS PAGE TO A FRIEND
Cryobiology
Effect of trehalose as an additive to dimethyl sulfoxide solutions on ice formation, cellular viability, and metabolism.
PMID 28063960
Abstract
Cryopreservation is the only established method for long-term preservation of cells and cellular material. This technique involves preservation of cells and cellular components in the presence of cryoprotective agents (CPAs) at liquid nitrogen temperatures (-196 °C). The organic solvent dimethyl sulfoxide (Me