EMAIL THIS PAGE TO A FRIEND
International orthopaedics
A descriptive study of potential effect of anterior tibial translation, femoral tunnel and anterior cruciate ligament graft inclination on clinical outcome and degenerative changes.
PMID 28064350
Abstract
There is no evidence that anatomically correct anterior cruciate ligament reconstruction (ACLR) offers lower rate of degenerative changes development or that it would lead to a better outcome. The significance and understanding of the abnormal anterior tibial translation (ATT) in ACLR patients is yet to be established. Sixty subjects (40 patients at 5.9 years after ACLR, 20 healthy controls) underwent 3 T MRI. Quantitative cartilage T2 mapping and morphological whole organ magnetic resonance imaging score (WORMS) evaluation was performed. Self-reported questionnaires were used for subjective clinical evaluation. Correlations were calculated with the following MRI measurements; femoral tunnel inclination, ACL graft inclination, lateral and medial compartment ATT. In the ACLR group positive correlation was found between the patellar cartilage T2 values and sagittal ACL graft inclination. In the ACLR group lateral compartment ATT showed negative correlation with ACL graft inclination and subjective clinical evaluation, and positive correlation with morphological degenerative changes. Femoral tunnel showed positive correlation with ACL graft inclination in the same plane. Increased ATT offers worse clinical outcome and increased rate of degenerative changes. Furthermore, ATT is affected by the ACL inclination. Inclination of the drilling tunnel affects ACL graft inclination; thereby independent drilling techniques provide superior results of anatomical ACL graft positioning.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
719536
3,3′-Diiodo-L-thyronine (T2), 98% (CP)
C15H13I2NO4