EMAIL THIS PAGE TO A FRIEND
Nature communications
The transcription factor EPAS1 links DOCK8 deficiency to atopic skin inflammation via IL-31 induction.
PMID 28067314
Abstract
Mutations of DOCK8 in humans cause a combined immunodeficiency characterized by atopic dermatitis with high serum IgE levels. However, the molecular link between DOCK8 deficiency and atopic skin inflammation is unknown. Here we show that CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8807
FM19G11, ≥98% (HPLC)
C23H17N3O8