EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Unusual N-oxide formation in the peroxidation of cobalt(ii) ethylenediaminetetraacetates.
PMID 28067375
Abstract
Unlike the usual formations of peroxo and superoxo cobalt products as oxygen carriers, N-oxido cobalt(ii) ethylenediaminetetraacetates K
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100277
Adamantane, ≥99%
C10H16