EMAIL THIS PAGE TO A FRIEND
Journal of breath research
A collection system for dry solid residues from exhaled breath for analysis via atomic force microscopy.
PMID 28068290
Abstract
Exhaled air contains sub-micron droplets of lung liquid, which potentially bear biomarkers of lung diseases. After dehydration they form dry residue particles (DRPs). As a first step in developing techniques to characterize individual DRPs, a new electrostatic collector was designed in which DRPs are charged within a unipolar corona charger, concentrated in a cone funnel, and deposited onto a limited area of a highly oriented pyrolytic graphite surface. The collector captures 80%-90% of DRPs at an optimal flow rate of 0.15 l min
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1348
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
C37H74NO8P