EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Influence of neuropeptide Y and pancreatic polypeptide on islet function and beta-cell survival.
PMID 28069397
Abstract
In the present study we assessed the impact of neuropeptide Y receptor (NPYR) modulators, neuropeptide Y (NPY) and pancreatic polypeptide (PP), on islet function and beta-cell survival. The effects of NPY and PP on beta-cell function were examined in BRIN BD11 and 1.1B4 beta-cells, as well as isolated mouse islets. Involvement of both peptides in pancreatic islet adaptations to streptozotocin and hydrocortisone, as well as effects on beta-cell proliferation and apoptosis was also evaluated. Neither NPY nor PP affected in vivo glucose disposal or insulin secretion in mice. However, both peptides inhibited (p<0.05 to p<0.001) glucose stimulated insulin secretion from rat and human beta-cells. NPY exerted similar insulinostatic effects in isolated mouse islets. NPY and PP inhibited alanine-induced changes in BRIN BD11 cell membrane potential and (Ca These data emphasise the involvement of PP, and particularly NPY, in the regulation of beta-cell mass and function. Modulation of PP and NPY signalling is suitable for further evaluation and possible clinical development for the treatment of diabetes.