EMAIL THIS PAGE TO A FRIEND
Advances in therapy
Pharmacokinetic Evaluation of Once-Weekly and Once-Monthly Buprenorphine Subcutaneous Injection Depots (CAM2038) Versus Intravenous and Sublingual Buprenorphine in Healthy Volunteers Under Naltrexone Blockade: An Open-Label Phase 1 Study.
PMID 28070862
Abstract
CAM2038 q1w (once weekly) and q4w (once monthly) are investigational buprenorphine subcutaneous (SC) formulations based on FluidCrystal Healthy volunteers were randomized to five treatment groups. All received a single intravenous dose of buprenorphine 600 µg, followed post-washout by a single dose of CAM2038 q4w 96 mg, a single dose of CAM2038 q4w 192 mg, or sublingual buprenorphine 8, 16, or 24 mg daily for 7 days, followed post-washout by a single dose of CAM2038 q4w 64 or 128 mg or four repeated weekly doses of CAM2038 q1w 16 mg. All subjects received daily naltrexone. Eighty-seven subjects were randomized. Median buprenorphine t The pharmacokinetic profiles of CAM2038 q1w and q4w versus sublingual buprenorphine support expected treatment efficacy with once-weekly and once-monthly dosing, respectively. CAM2038 formulations were safe and showed good local tolerability. ISRCTN24987553. Camurus AB.