EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Single Conformation Spectroscopy of Suberoylanilide Hydroxamic Acid: A Molecule Bites Its Tail.
PMID 28071906
Abstract
Suberoylanilide hydroxamic acid (SAHA) is a histone deacetylase inhibitor that causes growth arrest and differentiation of many tumor types and is an approved drug for the treatment of cancer. The chemical structure of SAHA consists of formanilide "head" and a hydroxamic acid "tail" separated by an n-hexyl chain, C