EMAIL THIS PAGE TO A FRIEND
American journal of translational research
Potential herb-drug interaction of shexiang baoxin pill in vitro based on drug metabolism/transporter.
PMID 28078025
Abstract
Many researches have proved functions of anti-oxidation, endothelial protection and pro-angiogenesis efficiency of Shexiang Baoxin Pill (SBP). This study aims to investigate potential for metabolism-based interaction on CYP450s and transporter based interaction on OATP1B1, BRCP and MDR1. Human primary hepatocytes were used in this study. Probe substrates of cytochrome P450 enzymes were incubated in human liver microsomes (HLMs) with or without SBP and IC50 values were estimated. Inhibitive potential of SBP on activities of CYP1A2, 2B6, 2C8, 2C9, 2C19, 2D6 and 3A4 was evaluated. Inducible potential of SBP on activities of CYP1A2, 2B6 and 3A4 was accessed. Inhibitive potential of SBP on human OATP1B1 was evaluated using cell-based assay. Inhibitive potential of SBP on human MDR1 and BCRP was also evaluated using vesicles assay. MDR1 and BCRP vesicle kit were used to determine ATP dependent uptake activity when incubated with SBP. SBP was a competitive inhibitor of CYP2B6, 2C19, while neither inhibitory nor inductive potentials toward other CYP450s were detected. No significant MDR1 inhibitory potential was estimated, while only high concentration of SBP (500 μg/ml) could inhibit activity of BCRP. Probe substrates Estradiol-17 β-glucuronide was incubated in HEK293-OATP1B1 and HEK293-MOCK cell system with different concentration of SBP and estimated IC50 was 179 μg/mL, which demonstrated a moderate inhibition potential against OATP1B1. In conclusion, outcome of this study suggests that SBP plays an important role in inhibition of CYP450 isozymes (including CYP2B6 and 2C9) and transporter OATP1B1. Therefore, precautions should be taken when using SBP for CYP and OATP-related herb-drug interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
UC205
Dextrorphan
C17H23NO
SBNMQ NMQ/N-methyl-quinidine - MDR1/P-gp Substrate
C21H27IN2O2