EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Covalent organic framework-coated magnetic graphene as a novel support for trypsin immobilization.
PMID 28078417
Abstract
Deep and efficient proteolysis is the critical premise in mass spectrometry-based bottom-up proteomics. It is difficult for traditional in-solution digestion to meet the requirement unless prolonged digestion time and enhanced enzyme dosage are employed, which makes the whole workflow time-consuming and costly. The abovementioned problems could be effectively ameliorated by anchoring many proteases on solid supports. In this work, covalent organic framework-coated magnetic graphene (MG@TpPa-1) was designed and prepared as a novel enzyme carrier for the covalent immobilization of trypsin with a high degree of loading (up to 268 μg mg