EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
In situ morphological assessment of apoptosis induced by Phaleria macrocarpa (Boerl.) fruit ethyl acetate fraction (PMEAF) in MDA-MB-231 cells by microscopy observation.
PMID 28081471
Abstract
Phaleria macrocarpa (Boerl.) is a well-known medicinal plant and have been extensively used as traditional medicine for ages in treatment of various diseases. The purpose of this study was to determine the in situ cytotoxicity effect P. macrocarpa fruit ethyl acetate fraction (PMEAF) by using various conventional and modern microscopy techniques. The cytotoxicity of PMEAF treated MDA-MB-231 cells was determined through the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) cytotoxicity assay and CyQuant Cell Proliferation Assay after 24h of treatment. Both results were indicated that the PMEAF is a potential anticancer agent with the average IC