EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Secretin is involved in sodium conservation through the renin-angiotensin-aldosterone system.
PMID 28082350
Abstract
Secretin (SCT) and its receptor (SCTR) are important in fluid regulation at multiple levels
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9477
Aldosterone, ≥95% (HPLC)
C21H28O5