EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
FTY720/fingolimod increases NPC1 and NPC2 expression and reduces cholesterol and sphingolipid accumulation in Niemann-Pick type C mutant fibroblasts.
PMID 28082351
Abstract
Niemann-Pick type C (NPC) disease is a fatal neurodegenerative disorder caused by mutations in