EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Identification of the metabolic defect in sialuria.
PMID 2808337
Abstract
Sialuria is a rare inborn error of metabolism, the hallmarks of which are moderate developmental retardation, coarse facial features, and an enormous amount of free N-acetylneuraminic acid (sialic acid) in the urine. Until now, the basic biochemical defect in this disorder has remained uncertain. In this report, the activity of the rate-limiting enzyme in the biosynthesis of sialic acid has been measured directly in whole cell lysates by a highly sensitive assay. With this technique, the basic defect in sialuria has been identified unequivocally as the loss of feedback control of uridine diphosphate N-acetylglucosamine 2-epimerase by cytidine monophosphate N-acetylneuraminic acid with resultant overproduction of sialic acid.