Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia
Co-expression of tissue factor and IL-6 in immature endothelial cells of cerebral cavernous malformations.
PMID 28087183
Cerebral cavernous malformations (CCMs) are congenital abnormal clusters of capillaries that are prone to leaking and thought to result from a disorder of endothelial cells. The underlying pathology of CCM is not fully understood. We analyzed the expression of tissue factor (TF) and interleukin-6 (IL-6) in CCMs to determine the association of TF and IL-6 with clinical and pathological findings. Thirteen cases of operative specimens of sporadic CCMs were included in this study. The expression of messenger RNA of TF and IL-6 was assayed and the association with clinical factors was investigated. Then, the distribution of TF and IL-6 was examined with immunofluorescence. The mRNA expression of TF of CCMs was significantly higher than that of the control (p=0.017), and was correlated with the number of hemorrhage appearances (p=0.190, ρ=0.62). The mRNA expression level of IL-6 was significantly correlated with the mRNA expression level of TF (p=0.034, ρ=0.58). Examination of immunostained sections indicated that TF+ cells were also positive for IL-6, and distributed around normal endothelial cells. Moreover, the TF+/IL-6+ cells expressed CD31 and VEGFR2. The expressions of IL-6 and TF were correlated, and both were present in the same immature endothelial cells. TF is elevated in CCM and might mediate progressive events. These factors may play a prognostic role in CCM.