EMAIL THIS PAGE TO A FRIEND
Cancer research
Sunitinib Stimulates Expression of VEGFC by Tumor Cells and Promotes Lymphangiogenesis in Clear Cell Renal Cell Carcinomas.
PMID 28087600
Abstract
Sunitinib is an antiangiogenic therapy given as a first-line treatment for renal cell carcinoma (RCC). While treatment improves progression-free survival, most patients relapse. We hypothesized that patient relapse can stem from the development of a lymphatic network driven by the production of the main growth factor for lymphatic endothelial cells, VEGFC. In this study, we found that sunitinib can stimulate
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9760
L-(−)-Dithiothreitol, ≥95%
C4H10O2S2