EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Topical corticosteroid compounding: effects on physicochemical stability and skin penetration rate.
PMID 2808834
Abstract
Compounding compatibility studies of four corticosteroid cream products and four commonly added chemicals are presented. Physical alteration, chemical stability, micropreservative challenge status, and in vitro skin penetration were evaluated at ambient conditions for 2 months. The study was designed to generate useful, previously unavailable information to aid dispensing pharmacists and dermatologists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5395
Hydrocortisone 17-valerate, analytical standard
C26H38O6