EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Simultaneous quantitation of sphingoid bases by UPLC-ESI-MS/MS with identical
PMID 28089753
Abstract
Free sphingoid bases (lysosphingolipids) of primary storage sphingolipids are increased in tissues and plasma of several sphingolipidoses. As shown earlier by us, sphingoid bases can be accurately quantified using UPLC-ESI-MS/MS, particularly in combination with identical
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
18448
N-Stearoyl-D-sphingosine, ≥98.0% (TLC)
C36H71NO3